Specifications
CAS13007-90-4
Chemical Name or MaterialBis(triphenylphosphine)dicarbonylnickel
DOT InformationTransport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
EINECS Number235-851-9
Formula Weight639.29
View more