Lanthanum aluminum oxide, 99.9% (metals basis)
Lanthanum aluminum oxide, 99.9% (metals basis)
Lanthanum aluminum oxide, 99.9% (metals basis)
Thermo Scientific Chemicals

Lanthanum aluminum oxide, 99.9% (metals basis)

CAS: 12003-65-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 039250.06
also known as 039250-06
Price (USD)
57.65
Special offer
Online exclusive
Ends: 28-Dec-2026
67.80
Save 10.15
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
57.65
Special offer
Online exclusive
Ends: 28-Dec-2026
67.80
Save 10.15
Each
Chemical Identifiers
CAS12003-65-5
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Used in epitaxial thin films, non-epitaxial thin films and substrate. Mainly used in making optical glass parts and optical fibers for electronic industry. Also often used in ceramic industry and catalysts. Due to its dielectric properties it is also posses properties for low-loss microwave and dielectric resonance applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only