Iron(II) phthalocyanine, 96%
Iron(II) phthalocyanine, 96%
Iron(II) phthalocyanine, 96%
Thermo Scientific Chemicals

Iron(II) phthalocyanine, 96%

CAS: 132-16-1 | C32H16FeN8 | 568.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 039262.03
also known as 039262-03
Price (USD)
37.65
Special offer
Online exclusive
Ends: 30-Jun-2026
44.40
Save 6.75 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
37.65
Special offer
Online exclusive
Ends: 30-Jun-2026
44.40
Save 6.75 (15%)
Each
Chemical Identifiers
CAS132-16-1
IUPAC Nameλ²-iron(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide
Molecular FormulaC32H16FeN8
InChI KeyMIINHRNQLVVCEW-UHFFFAOYSA-N
SMILES[Fe++].[N-]1C2=NC3=NC(=NC4=C5C=CC=CC5=C([N-]4)N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark purple
Formcrystalline powder
Assay (Titration)> 95.0%
Iron phthalocyanine is been investigated as organic electroluminescence materials for the applications in organic solar cells, biosensitizers and display devices such as OLED(Organic Light Emiting Diode), OTFT(Organic Thin Film Transistor), Wearable Display, and e-paper. It is also used in the formation of new type of catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only