Iron(III) 1,1,1-trifluoro 2,4-pentanedionate
Iron(III) 1,1,1-trifluoro 2,4-pentanedionate
Iron(III) 1,1,1-trifluoro 2,4-pentanedionate
Thermo Scientific Chemicals

Iron(III) 1,1,1-trifluoro 2,4-pentanedionate

CAS: 14526-22-8 | C15H12F9FeO6 | 515.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 039265.14
also known as 039265-14
Price (USD)
545.65
Online Exclusive
606.00
Save 60.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
545.65
Online Exclusive
606.00
Save 60.35 (10%)
Each
Specifications
CAS14526-22-8
Chemical Name or MaterialIron(III) 1,1,1-trifluoro 2,4-pentanedionate
Formula Weight515.09
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
It is used as an intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only