Pentamethylcyclopentadienyliron dicarbonyl dimer
Pentamethylcyclopentadienyliron dicarbonyl dimer
Pentamethylcyclopentadienyliron dicarbonyl dimer
Thermo Scientific Chemicals

Pentamethylcyclopentadienyliron dicarbonyl dimer

CAS: 35344-11-7 | C24H30Fe2O4 | 494.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 039266.06
also known as 039266-06
Price (USD)
451.65
Online Exclusive
502.00
Save 50.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
451.65
Online Exclusive
502.00
Save 50.35 (10%)
Each
Specifications
CAS35344-11-7
Chemical Name or MaterialPentamethylcyclopentadienyliron dicarbonyl dimer
ColorPurple-Red
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
Formula Weight493.96
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only