1,2-Bis(dicyclohexylphosphino)ethane, 98%
1,2-Bis(dicyclohexylphosphino)ethane, 98%
1,2-Bis(dicyclohexylphosphino)ethane, 98%
Thermo Scientific Chemicals

1,2-Bis(dicyclohexylphosphino)ethane, 98%

CAS: 23743-26-2 | C26H48P2 | 422.618 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
Catalog number 039269.04
also known as 039269-04
Price (USD)
314.65
Special offer
Online exclusive
Ends: 30-Jun-2026
370.00
Save 55.35 (15%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
314.65
Special offer
Online exclusive
Ends: 30-Jun-2026
370.00
Save 55.35 (15%)
Each
Specifications
CAS23743-26-2
Chemical Name or Material1,2-Bis(dicyclohexylphosphino)ethane
DOT InformationTransport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
Formula Weight422.61
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only