Bis(dimethylamino)phosphorochloridate, 94%, Thermo Scientific Chemicals
Bis(dimethylamino)phosphorochloridate, 94%, Thermo Scientific Chemicals
Bis(dimethylamino)phosphorochloridate, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(dimethylamino)phosphorochloridate, 94%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
039274.04
also known as 039274-04
2 g
Catalog number 039274.04
also known as 039274-04
Price (USD)
42.60
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
42.60
Each
Specifications
Beilstein471430
CAS1605-65-8
Chemical Name or MaterialBis(dimethylamino)phosphorochloridate
ColorColorless
DOT InformationTransport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only