Bis(ethylene)(2,4-pentanedionato)rhodium(I), Rh 39.9% min
Bis(ethylene)(2,4-pentanedionato)rhodium(I), Rh 39.9% min
Bis(ethylene)(2,4-pentanedionato)rhodium(I), Rh 39.9% min
Thermo Scientific Chemicals

Bis(ethylene)(2,4-pentanedionato)rhodium(I), Rh 39.9% min

CAS: 12082-47-2 | C9H15O2Rh | 258.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 039288.03
also known as 039288-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS12082-47-2
IUPAC Nameλ¹-rhodium(1+) bis(ethene) (2Z)-4-oxopent-2-en-2-olate
Molecular FormulaC9H15O2Rh
InChI KeyFLRBEQQDEGBCJS-FGSKAQBVSA-M
SMILES[Rh+].C=C.C=C.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Rh 39.9% min
It acts as a hydrogenation precursor. It is also used as intermediates which acts as catalyst such as in olefin hydroformylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only