Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer, 99%
Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer, 99%
Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer, 99%
Thermo Scientific Chemicals

Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer, 99%

CAS: 12132-04-6 | C12H16MoO2 | 288.208 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 039303.03
also known as 039303-03
Price (USD)
126.65
Special offer
Online exclusive
Ends: 30-Jun-2026
149.00
Save 22.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
126.65
Special offer
Online exclusive
Ends: 30-Jun-2026
149.00
Save 22.35 (15%)
Each
Specifications
CAS12132-04-6
Chemical Name or MaterialPentamethylcyclopentadienylmolybdenum dicarbonyl dimer
DOT InformationTransport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
Formula Weight574.38
Health Hazard 1H300+H330
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only