Manganese(II) phthalocyanine
Manganese(II) phthalocyanine
Manganese(II) phthalocyanine
Thermo Scientific Chemicals

Manganese(II) phthalocyanine

CAS: 14325-24-7 | C32H16MnN8 | 567.474 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 039309.06
also known as 039309-06
Price (USD)
171.65
Online Exclusive
191.00
Save 19.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
171.65
Online Exclusive
191.00
Save 19.35 (10%)
Each
Specifications
CAS14325-24-7
Chemical Name or MaterialManganese(II) phthalocyanine
Formula Weight567.47
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only