Magnesium meso-tetraphenylporphine monohydrate
Magnesium meso-tetraphenylporphine monohydrate
Magnesium meso-tetraphenylporphine monohydrate
Thermo Scientific Chemicals

Magnesium meso-tetraphenylporphine monohydrate

CAS: 14640-21-2 | C44H28MgN4 | 637.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
Catalog number 039316.02
also known as 039316-02
Price (USD)
130.65
Special offer
Online exclusive
Ends: 30-Jun-2026
153.00
Save 22.35 (15%)
Each
Quantity:
0.5 g
Request bulk or custom format
Price (USD)
130.65
Special offer
Online exclusive
Ends: 30-Jun-2026
153.00
Save 22.35 (15%)
Each
Specifications
CAS14640-21-2
Chemical Name or MaterialMagnesium meso-tetraphenylporphine monohydrate
Formula Weight655.05 (637.04 Anhydrous)
IUPAC Namemagnesium;2,7,12,17-tetraphenylporphyrin-21,22-diide
InChI KeyXEHJAWQTIIXDON-QHNUXLDVSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only