Magnesium pyrophosphate
Magnesium pyrophosphate
Magnesium pyrophosphate
Magnesium pyrophosphate
Thermo Scientific Chemicals

Magnesium pyrophosphate

CAS: 13446-24-7 | Mg2O7P2 | 222.551 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 039317.30
also known as 039317-30
Price (USD)
178.65
Online Exclusive
198.00
Save 19.35 (10%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
178.65
Online Exclusive
198.00
Save 19.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS13446-24-7
IUPAC Namedimagnesium(2+) (phosphonatooxy)phosphonate
Molecular FormulaMg2O7P2
InChI KeyXZTWHWHGBBCSMX-UHFFFAOYSA-J
SMILES[Mg++].[Mg++].[O-]P([O-])(=O)OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)21% Mg min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only