Aluminum magnesium isopropoxide, 99.95+% (metals basis)
Thermo Scientific Chemicals

Aluminum magnesium isopropoxide, 99.95+% (metals basis)

CAS: 69207-83-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 039320.18
also known as 039320-18
Price (USD)
737.65
Online Exclusive
819.00
Save 81.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
737.65
Online Exclusive
819.00
Save 81.35 (10%)
Each
Specifications
Assay Percent Notes(metals basis)
CAS69207-83-6
Chemical Name or MaterialAluminum magnesium isopropoxide
DOT InformationTransport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
Formula Weight550.53
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only