Lithium trifluoromethanesulfonate, 97%
Lithium trifluoromethanesulfonate, 97%
Lithium trifluoromethanesulfonate, 97%
Thermo Scientific Chemicals

Lithium trifluoromethanesulfonate, 97%

CAS: 33454-82-9 | CF3LiO3S | 156.003 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 039322.18
also known as 039322-18
Price (USD)
153.65
Online Exclusive
171.00
Save 17.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
153.65
Online Exclusive
171.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS33454-82-9
IUPAC Nametrifluoromethanesulfonic acid lithium
Molecular FormulaCHF3LiO3S
InChI KeyWDGKXRCNMKPDSD-UHFFFAOYSA-N
SMILES[Li].OS(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey
FormPowder
Assay (unspecified)Lithium: ≥4.27% (theoretical: 4.45%)
Lithium trifluoromethanesulfonate is mixed with ceramic filler to prepare polyethylene oxide films in order to improve the electrochemical and mechanical characteristics of the membrane, which is used in the lithium batteries. It acts as a doping salt used in the preparation of nano manganese-composite polymer electrolytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only