Lead(II) metaborate monohydrate, Thermo Scientific Chemicals
Lead(II) metaborate monohydrate, Thermo Scientific Chemicals
Lead(II) metaborate monohydrate, Thermo Scientific Chemicals
Lead(II) metaborate monohydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead(II) metaborate monohydrate, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
039339.22
also known as 039339-22
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 039339.22
also known as 039339-22
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS10214-39-8
IUPAC Nameλ²-lead(2+) hydrate dioxoborinate
Molecular FormulaB2H2O5Pb
InChI KeyRTSNZWSJUUUHCZ-UHFFFAOYSA-N
SMILESO.[Pb++].[O-]B=O.[O-]B=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98% min
Lead(II) metaborate monohydrate acts as a starting material used in the preparation of lead(II) borates such as Pb2B3O5.5 (OH)2 and [Pb3(B3O7)](NO3) by changing the pH values of the reaction systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only