Product Image
Thermo Scientific™
Dionex™ Postcolumn Reaction Coils (Pneumatic or Pump Delivery)
The Thermo Scientific™ Dionex™ Postcolumn Reaction Coils (Pneumatic or Pump Delivery) are for use with the Detector/Chromatography (DC) Compartment.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity
039349500 μL
037859750 μL
Catalog number 039349
Price (MXN)
-
Capacity:
500 μL
The Thermo Scientific™ Dionex™ Postcolumn Reaction Coils (Pneumatic or Pump Delivery) are for use with the Detector/Chromatography (DC) Compartment.
  • 5°C above DC upper compartment temperature, to 80°C
  • 2 coils
  • For PCH/RCH
Specifications
Capacity500 μL
DescriptionKnitted Reaction Coil, 500 μL, Potted (for PCH/RCH)
For Use With (Equipment)Detector/Chromatography (DC) Compartment
Includes2 coils
Temperature Range5°C above DC upper compartment temperature, to 80°C
TypeReaction Coil
Unit SizeEach