Lithium salicylate, tech.
Lithium salicylate, tech.
Lithium salicylate, tech.
Thermo Scientific Chemicals

Lithium salicylate, tech.

CAS: 552-38-5 | C7H5LiO3 | 144.054 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 039356.22
also known as 039356-22
Price (USD)
97.90
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
97.90
Each
Specifications
CAS552-38-5
Chemical Name or MaterialLithium salicylate
EINECS Number209-011-7
Formula Weight144.05
Health Hazard 1H302-H314-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only