Lithium tartrate monohydrate, 99%
Lithium tartrate monohydrate, 99%
Lithium tartrate monohydrate, 99%
Thermo Scientific Chemicals

Lithium tartrate monohydrate, 99%

CAS: 6108-32-3 | C4H6Li2O7 | 179.965 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 039357.18
also known as 039357-18
Price (USD)
59.90
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
59.90
Each
Specifications
CAS6108-32-3
Chemical Name or MaterialLithium tartrate monohydrate
EINECS Number212-772-8
Formula Weight179.97 (161.97 Anhydrous)
IUPAC Namedilithium;2,3-dihydroxybutanedioate;hydrate
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only