Lithium vanadium oxide, 99.9% (metals basis)
Lithium vanadium oxide, 99.9% (metals basis)
Lithium vanadium oxide, 99.9% (metals basis)
Thermo Scientific Chemicals

Lithium vanadium oxide, 99.9% (metals basis)

CAS: 15060-59-0
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 039358.30
also known as 039358-30
Price (USD)
315.65
Special offer
Online exclusive
Ends: 30-Jun-2026
371.00
Save 55.35 (15%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
315.65
Special offer
Online exclusive
Ends: 30-Jun-2026
371.00
Save 55.35 (15%)
Each
Specifications
Assay Percent Notes(metals basis)
CAS15060-59-0
Chemical Name or MaterialLithium vanadium oxide
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.
EINECS Number239-124-7
View more
Lithium vanadium oxide is used in developing polymer lithium ion (PLI) batteries and also in coatings, nanowires, nanofibers, plastics, and textiles In alloy and catalyst applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only