Lithium isobutyrate, 98+%
Lithium isobutyrate, 98+%
Lithium isobutyrate, 98+%
Thermo Scientific Chemicals

Lithium isobutyrate, 98+%

CAS: 25179-23-1 | C4H7LiO2 | 94.038 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Catalog number 039372.36
also known as 039372-36
Price (USD)
455.00
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
455.00
Each
Chemical Identifiers
CAS25179-23-1
IUPAC Namelithium(1+) 2-methylpropanoate
Molecular FormulaC4H7LiO2
InChI KeyIIDVGIFOWJJSIJ-UHFFFAOYSA-M
SMILES[Li+].CC(C)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or lump or solid
Assay (unspecified)Lithium (Li): 7.19-7.56% (Therory: 7.38%)
Assay from Suppliers CofA≥97.5%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only