Platinum Iridium wire, 0.25mm (0.01in) dia, annealed, 99.9+% (metals basis)
Platinum Iridium wire, 0.25mm (0.01in) dia, annealed, 99.9+% (metals basis)
Platinum Iridium wire, 0.25mm (0.01in) dia, annealed, 99.9+% (metals basis)
Thermo Scientific Chemicals

Platinum Iridium wire, 0.25mm (0.01in) dia, annealed, 99.9+% (metals basis)

Form: approx. 0.11g/10cm,; Temper: soft | CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 cm
50 cm
Catalog number 039383.BS
also known as 039383-BS
Price (USD)
-
Quantity:
10 cm
Request bulk or custom format
Specifications
AppearanceSilver gray wire
Assay Percent Range>99.9%
CAS6-4-7440
ChEBICHEBI:33400
Chemical Name or MaterialPlatinum Iridium
View more
Used in the fabrication of metal microelectrodes for electrical stimulation of nervous tissues and in electrophysiological recordings. Inoculating needles and leadwires for implants. Used in electrode materials, composing the gel boxes/electrophoresis chambers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only