Indium(I) bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals
Indium(I) bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals
Indium(I) bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals
Indium(I) bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Indium(I) bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
039388.14
also known as 039388-14
25 g
Catalog number 039388.14
also known as 039388-14
Price (USD)
915.65
Online Exclusive
1,017.00
Save 101.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
915.65
Online Exclusive
1,017.00
Save 101.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS14280-53-6
IUPAC Nameindium(3+) tribromide
Molecular FormulaBr3In
InChI KeyJKNHZOAONLKYQL-UHFFFAOYSA-K
SMILES[Br-].[Br-].[Br-].[In+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size-10 Mesh
FormBeads
Residual water< 100 ppm
Total Metal Impurities0.002% max.
Appearance (Color)Red
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only