tert-Butylferrocene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

tert-Butylferrocene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 039405.06
also known as 039405-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Specifications
CAS1316-98-9
Chemical Name or Materialtert-Butylferrocene
Density1.201 g/mL
Formula Weight242.15
MDL NumberMFCD00058729
View more
It finds its application in microwave-enhanced electrochemical processes in micellar surfactant media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only