Dicarbonyl(2,4-pentanedionato)iridium(I)
Dicarbonyl(2,4-pentanedionato)iridium(I)
Dicarbonyl(2,4-pentanedionato)iridium(I)
Thermo Scientific Chemicals

Dicarbonyl(2,4-pentanedionato)iridium(I)

CAS: 14023-80-4 | C7H7IrO4 | 347.35 g/mol
Have Questions?
Catalog NumberQuantity
039411.02
also known as 039411-02
0.5 g
Catalog number 039411.02
also known as 039411-02
Price (USD)
-
Quantity:
0.5 g
Request bulk or custom format
Chemical Identifiers
CAS14023-80-4
IUPAC Nameiridium(1+) bis(methanidylidyneoxidanium) (2Z)-4-oxopent-2-en-2-olate
Molecular FormulaC7H7IrO4
InChI KeyDFIAYCFHPDOZLK-FGSKAQBVSA-M
SMILES[Ir+].[C-]#[O+].[C-]#[O+].C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only