Terbium(III) bromide, ultra dry, 99.99% (REO)
Terbium(III) bromide, ultra dry, 99.99% (REO)
Terbium(III) bromide, ultra dry, 99.99% (REO)
Thermo Scientific Chemicals

Terbium(III) bromide, ultra dry, 99.99% (REO)

CAS: 14456-47-4 | Br3Tb | 398.637 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 039425.06
also known as 039425-06
Price (USD)
565.65
Online Exclusive
628.00
Save 62.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
565.65
Online Exclusive
628.00
Save 62.35 (10%)
Each
Chemical Identifiers
CAS14456-47-4
IUPAC Nameterbium(3+) tribromide
Molecular FormulaBr3Tb
InChI KeyAZNZWHYYEIQIOC-UHFFFAOYSA-K
SMILES[Br-].[Br-].[Br-].[Tb+3]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Residual water< 100 ppm
Appearance (Color)White
Total Rare Earth Oxide Impurities0.01% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only