Nickel(II) phthalocyanine
Nickel(II) phthalocyanine
Nickel(II) phthalocyanine
Thermo Scientific Chemicals

Nickel(II) phthalocyanine

CAS: 14055-02-8 | C32H16N8Ni | 571.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 039453.06
also known as 039453-06
Price (USD)
66.40
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
66.40
Each
Chemical Identifiers
CAS14055-02-8
IUPAC Namenickel(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide
Molecular FormulaC32H16N8Ni
InChI KeyLVIYYTJTOKJJOC-UHFFFAOYSA-N
SMILES[Ni++].[N-]1C2=NC3=NC(=NC4=C5C=CC=CC5=C([N-]4)N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥95.0%
FormPowder
Water Content≤1.0% (loss on drying)
Appearance (Color)Dark blue
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only