Nickel(II) octanoate, in mineral spirits (8% Ni)
Nickel(II) octanoate, in mineral spirits (8% Ni)
Nickel(II) octanoate, in mineral spirits (8% Ni)
Thermo Scientific Chemicals

Nickel(II) octanoate, in mineral spirits (8% Ni)

CAS: 4995-91-9 | C8H16NiO2 | 202.907 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 039479.36
also known as 039479-36
Price (USD)
463.65
Online Exclusive
515.00
Save 51.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
463.65
Online Exclusive
515.00
Save 51.35 (10%)
Each
Specifications
CAS4995-91-9
Chemical Name or MaterialNickel(II) octanoate
ColorGreen
Concentration or Composition (by Analyte or Components)8% Ni
DOT InformationTransport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
View more
Nickel(II) octanoate is used to prepare nickel based catalysts using niobia modified silica as support. It is also used as a catalyst and involved in the treatment of qaiyarah heavy crude oil, petrolene using ethanol as a solvent and reductant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only