Palladium hydroxide, 99%
Palladium hydroxide, 99%
Palladium hydroxide, 99%
Thermo Scientific Chemicals

Palladium hydroxide, 99%

CAS: 12135-22-7 | H2O2Pd | 140.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.25 g
1 g
Catalog number 039496.01
also known as 039496-01
Price (USD)
79.65
Special offer
Online exclusive
Ends: 30-Jun-2026
94.10
Save 14.45 (15%)
Each
Quantity:
0.25 g
Request bulk or custom format
Price (USD)
79.65
Special offer
Online exclusive
Ends: 30-Jun-2026
94.10
Save 14.45 (15%)
Each
Chemical Identifiers
CAS12135-22-7
IUPAC Namepalladium(2+) dihydroxide
Molecular FormulaH2O2Pd
InChI KeyNXJCBFBQEVOTOW-UHFFFAOYSA-L
SMILES[OH-].[OH-].[Pd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min
Acts as a catalyst for arylation reactions. It is employed in hydrogenolysis of benzyl-nitrogen bonds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only