Zinc meso-tetraphenylporphine
Zinc meso-tetraphenylporphine
Zinc meso-tetraphenylporphine
Thermo Scientific Chemicals

Zinc meso-tetraphenylporphine

CAS: 14074-80-7 | C44H28N4Zn | 678.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
Catalog number 039552.02
also known as 039552-02
Price (USD)
93.65
Special offer
Online exclusive
Ends: 30-Jun-2026
110.00
Save 16.35 (15%)
Each
Quantity:
0.5 g
Request bulk or custom format
Price (USD)
93.65
Special offer
Online exclusive
Ends: 30-Jun-2026
110.00
Save 16.35 (15%)
Each
Specifications
CAS14074-80-7
Chemical Name or MaterialZinc meso-tetraphenylporphine
ColorPurple
Formula Weight678.1
IUPAC Name5,10,15,20-tetraphenylporphyrin-22,24-diide;zinc
View more
A porphyrin used in light emitting diodes. It has valuable biological applications in molecular biology, fluoroimmunoassay and new pharmaceutical developments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only