Zinc naphthenate, ^=67% in mineral spirits (10% Zn)
Zinc naphthenate, ^=67% in mineral spirits (10% Zn)
Zinc naphthenate, ^=67% in mineral spirits (10% Zn)
Thermo Scientific Chemicals

Zinc naphthenate, ^=67% in mineral spirits (10% Zn)

CAS: 12001-85-3
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 039554.36
also known as 039554-36
Price (USD)
84.65
Online Exclusive
94.20
Save 9.55 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
84.65
Online Exclusive
94.20
Save 9.55 (10%)
Each
Chemical Identifiers
CAS12001-85-3
IUPAC Namezinc(2+) bis(3-(3-ethylcyclopentyl)propanoate)
Molecular FormulaC20H34O4Zn
InChI KeyHNWHEAHEQDAIDC-UHFFFAOYNA-L
SMILES[Zn++].CCC1CCC(CCC([O-])=O)C1.CCC1CCC(CCC([O-])=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormViscous liquid
Assay from Supplier's CofAZn: 9.0-11.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only