Diethylzinc, 96%
Diethylzinc, 96%
Diethylzinc, 96%
Thermo Scientific Chemicals

Diethylzinc, 96%

CAS: 557-20-0 | C4H10Zn | 123.50 g/mol
Have Questions?
Catalog NumberQuantityPackaging
039564.22
also known as 039564-22
100 gCylinder
Catalog number 039564.22
also known as 039564-22
Price (USD)
283.65
Special offer
Online exclusive
Ends: 30-Jun-2026
333.00
Save 49.35 (15%)
Each
Quantity:
100 g
Packaging:
Cylinder
Request bulk or custom format
Price (USD)
283.65
Special offer
Online exclusive
Ends: 30-Jun-2026
333.00
Save 49.35 (15%)
Each
Chemical Identifiers
CAS557-20-0
IUPAC Namediethylzinc
Molecular FormulaC4H10Zn
InChI KeyHQWPLXHWEZZGKY-UHFFFAOYSA-N
SMILESCC[Zn]CC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥95.0%
FormLiquid
It is used as an important reagent in organic chemistry. It is used in organic synthesis as a source of the ethyl carbanion in addition reactions to carbonyl groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only