Dionex™ IonPac™ NG1-10μm Guard Column (4 x 35 mm)
Actual product may vary
Dionex™ IonPac™ NG1-10μm Guard Column (4 x 35 mm)
Dionex™ IonPac™ NG1-10μm Guard Column (4 x 35 mm)
Thermo Scientific™

Dionex™ IonPac™ NG1-10μm Guard Column (4 x 35 mm)

Green features
Column Format: Guard Column; Column Phase: Reversed Phase, Ion-Pairing, Ion-Suppression
Have Questions?
Catalog NumberCrosslinking
03956755%DVB
Catalog number 039567
Price (USD)
368.50
Each
Crosslinking:
55%DVB
Price (USD)
368.50
Each
Achieve high-efficiency separations of hydrophobic, ionizable compounds with Thermo Scientific™ Dionex™ IonPac™ NS1 10μm Analytical & Guard Columns. The columns of choice for ion-pair or ion-suppression chromatography, these polymer-based, reversed-phase columns provide excellent resolution and peak shape. Packing is a neutral, macroporous, high surface area, ethylvinylbenzene polymer cross linked with 55% divinylbenzene, making IonPac NS1 columns resistant to solvents, acids and bases while permitting the use of eluent from pH 0 to 14.
Polymer-Based Columns for Efficient Reversed-Phase Separations
  • Ideal for separation of large molecules that carry localized charges, such as surfactants
  • Can be used for traditional polymeric reversed-phase applications
  • Use with ion-pair chromatography for difficult separations
  • Available in 5 and 10μm particle sizes
Key Applications
Use IonPac NS1 columns for reliable separations of various compounds: High molecular weight aliphatic, carboxylic acids; Alkyl and aromatic sulfates and sulfonates; Quaternary ammonium ions; Water-soluble vitamins; Sulfur oxides; Metal cyanide complexes; Phenols
Specifications
Crosslinking55%DVB
Flow Rate1 mL/min
For Use With (Application)Reversed-Phase or Ion-Pair Chromatography
Green FeaturesFSC
HydrophobicityHigh
Max. Pressure4000 psi (275 bar)
Packing MaterialSpherical, Crosslinked, Macroporous Polymer
Particle Size10 μm
Stationary PhaseEthylvinylbenzene/Divinylbenzene
TypeMixed Mode
Column TypeGuard
Diameter (Metric)4 mm
FormatStandard Bore
Length (Metric)35 mm
pH0 to 14
Unit SizeEach