Dicarbonylbis(cyclopentadienyl)titanium(II), 98%
Dicarbonylbis(cyclopentadienyl)titanium(II), 98%
Dicarbonylbis(cyclopentadienyl)titanium(II), 98%
Thermo Scientific Chemicals

Dicarbonylbis(cyclopentadienyl)titanium(II), 98%

CAS: 12129-51-0 | C12H12O2Ti | 236.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
Catalog number 039571.02
also known as 039571-02
Price (USD)
103.65
Special offer
Online exclusive
Ends: 30-Jun-2026
122.00
Save 18.35 (15%)
Each
Quantity:
0.5 g
Request bulk or custom format
Price (USD)
103.65
Special offer
Online exclusive
Ends: 30-Jun-2026
122.00
Save 18.35 (15%)
Each
Specifications
CAS12129-51-0
Chemical Name or MaterialDicarbonylbis(cyclopentadienyl)titanium(II)
DOT InformationTransport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
Formula Weight234.09
Health Hazard 1H228-H301-H332
View more
It is used as an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only