Tris(methylcyclopentadienyl)yttrium(III), 99.9% (REO), Thermo Scientific Chemicals
Tris(methylcyclopentadienyl)yttrium(III), 99.9% (REO), Thermo Scientific Chemicals
Tris(methylcyclopentadienyl)yttrium(III), 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tris(methylcyclopentadienyl)yttrium(III), 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
039576.06
also known as 039576-06
5 g
Catalog number 039576.06
also known as 039576-06
Price (USD)
933.65
Online Exclusive
1,037.00
Save 103.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
933.65
Online Exclusive
1,037.00
Save 103.35 (10%)
Each
Specifications
Assay Percent Notes(REO)
CAS329735-72-0
Chemical Name or MaterialTris(methylcyclopentadienyl)yttrium(III)
ColorYellow
DOT InformationHazard Class: 4.1
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only