Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)ytterbium(III), 99%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)ytterbium(III), 99%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)ytterbium(III), 99%
Thermo Scientific Chemicals

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)ytterbium(III), 99%

CAS: 15492-52-1 | C33H57O6Yb | 722.867 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 039581.06
also known as 039581-06
Price (USD)
334.65
Online Exclusive
372.00
Save 37.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
334.65
Online Exclusive
372.00
Save 37.35 (10%)
Each
Chemical Identifiers
CAS15492-52-1
IUPAC Nameytterbium(3+) tris(2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate)
Molecular FormulaC33H57O6Yb
InChI KeyKACMHCBXMZUMQV-UHFFFAOYSA-K
SMILES[Yb+3].CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥98.5%
It is used as reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only