Vanadium(IV) oxide phthalocyanine
Vanadium(IV) oxide phthalocyanine
Vanadium(IV) oxide phthalocyanine
Thermo Scientific Chemicals

Vanadium(IV) oxide phthalocyanine

CAS: 13930-88-6 | C32H16N8OV | 579.476 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
10 g
Catalog number 039587.09
also known as 039587-09
Price (USD)
286.65
Online Exclusive
318.00
Save 31.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
286.65
Online Exclusive
318.00
Save 31.35 (10%)
Each
Chemical Identifiers
CAS13930-88-6
IUPAC Name2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19(39),20,22,24,26,28,30,32,34-nonadecaene-37,38-diide; oxovanadiumbis(ylium)
Molecular FormulaC32H16N8OV
InChI KeyYRZZLAGRKZIJJI-UHFFFAOYSA-N
SMILESO=[V++].[N-]1C2=NC3=C4C=CC=CC4=C([N-]3)N=C3N=C(N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark blue to purple
FormPowder
Assay from Suppliers CofA≥80.0%
Infrared spectrumConforms
It is used as an organic light-emitting diode intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only