Tetrachlorobis(tetrahydrofuran)titanium(IV), 98%, Thermo Scientific Chemicals
Tetrachlorobis(tetrahydrofuran)titanium(IV), 98%, Thermo Scientific Chemicals
Tetrachlorobis(tetrahydrofuran)titanium(IV), 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrachlorobis(tetrahydrofuran)titanium(IV), 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
039594.09
also known as 039594-09
10 g
Catalog number 039594.09
also known as 039594-09
Price (USD)
174.65
Online Exclusive
194.00
Save 19.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
174.65
Online Exclusive
194.00
Save 19.35 (10%)
Each
Chemical Identifiers
CAS31011-57-1
IUPAC Namebis(oxolane); tetrachlorotitanium
Molecular FormulaC8H16Cl4O2Ti
InChI KeyLXWBMENBONGPSB-UHFFFAOYSA-J
SMILESCl[Ti](Cl)(Cl)Cl.C1CCOC1.C1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Yellow
Assay (unspecified)>97.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only