Pentamethylcyclopentadienyltitanium trichloride
Pentamethylcyclopentadienyltitanium trichloride
Pentamethylcyclopentadienyltitanium trichloride
Thermo Scientific Chemicals

Pentamethylcyclopentadienyltitanium trichloride

CAS: 12129-06-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 039596.06
also known as 039596-06
Price (USD)
420.65
Online Exclusive
467.00
Save 46.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
420.65
Online Exclusive
467.00
Save 46.35 (10%)
Each
Specifications
CAS12129-06-5
Chemical Name or MaterialPentamethylcyclopentadienyltitanium trichloride
ColorRed
DOT InformationTransport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Formula Weight289.47
View more
It is used as a catalyst in Transesterification reactions and styrene polymerization. It is also used as Piano-stool catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only