Dionex™ MMS Regenerant Concentrates
Actual product may vary
Thermo Scientific™

Dionex™ MMS Regenerant Concentrates

Increase productivity and provide reliable analyses with Thermo Scientific™ MMS Regenerant Concentrates.
Catalog NumberVolume (Metric)
039602500 mL
0371644 x 200 mL
03960150 mL
039203200 mL
Catalog number 039602
Price (GBP)
127.00
Each
Volume (Metric):
500 mL
Price (GBP)
127.00
Each

Increase productivity and provide reliable analyses with Thermo Scientific™ MMS Regenerant Concentrates.

  • Formulated with high-purity, reagent-grade chemicals and water (18 MΩ-cm); quality-control checked for ionic contamination, are used
  • All standards are NIST™ traceable

Includes:

A Certificate of Analysis is provided with every standard and chelation chromatography reagent

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With (Application)HPLC
HazardsCorrosive
QuantityEach
Standard ComponentsTetrabutyl Ammonium Hydroxide Aqueous Solution
Standard Concentration2.6, 97.4%
StorageRoom Temperature
Volume (Metric)500 mL
TypeRegenerant Concentrate
Unit SizeEach