Tri-n-butyltin fluoride
Tri-n-butyltin fluoride
Tri-n-butyltin fluoride
Tri-n-butyltin fluoride
Thermo Scientific Chemicals

Tri-n-butyltin fluoride

CAS: 1983-10-4 | C8H15NaO2 | 166.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 039602.06
also known as 039602-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1983-10-4
IUPAC Nametributyltin fluoride
Molecular FormulaC12H29FSn
InChI KeySTUMXSORVXBBJR-UHFFFAOYSA-N
SMILESF.CCCC[SnH](CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
FormPowder
Tri-n-butyltin fluoride is a reagent used for the selective desilylation of bis(silyl) enol ethers. It is also used as antifoulant for marine paints; reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only