Tin(II) trifluoromethanesulfonate, 97%
Tin(II) trifluoromethanesulfonate, 97%
Tin(II) trifluoromethanesulfonate, 97%
Thermo Scientific Chemicals

Tin(II) trifluoromethanesulfonate, 97%

CAS: 62086-04-8 | CH4F3O3SSn+ | 271.805 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 039603.06
also known as 039603-06
Price (USD)
227.65
Online Exclusive
253.00
Save 25.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
227.65
Online Exclusive
253.00
Save 25.35 (10%)
Each
Specifications
CAS62086-04-8
Chemical Name or MaterialTin(II) trifluoromethanesulfonate
DOT InformationHazard Class: 8; Packaging Group: III
Formula Weight416.82
Health Hazard 1H314-H500
View more
Tin(II) trifluoromethanesulfonate is a mild Lewis acid used for stereoselective aldol and Michael reactions; [2,3]-Wittig and Ireland-Claisen rearrangements, Horner-Wadsworth-Emmons reactions, asymmetric [3 + 2] cycloaddition, ring opening reactions and Friedel-Crafts reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only