Di-tert-butyltin dichloride, 98%
Di-tert-butyltin dichloride, 98%
Di-tert-butyltin dichloride, 98%
Thermo Scientific Chemicals

Di-tert-butyltin dichloride, 98%

CAS: 19429-30-2 | C8H18Cl2Sn | 303.842 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
Catalog number 039619.02
also known as 039619-02
Price (USD)
43.80
Each
Quantity:
0.5 g
Request bulk or custom format
Price (USD)
43.80
Each
Specifications
CAS19429-30-2
Chemical Name or MaterialDi-tert-butyltin dichloride
ColorWhite
DOT InformationTransport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.
EINECS Number243-051-6
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only