Tantalum(V) tetraethoxide 2,4-pentanedionate, 99.99%, (metals basis)
Tantalum(V) tetraethoxide 2,4-pentanedionate, 99.99%, (metals basis)
Tantalum(V) tetraethoxide 2,4-pentanedionate, 99.99%, (metals basis)
Tantalum(V) tetraethoxide 2,4-pentanedionate, 99.99%, (metals basis)
Thermo Scientific Chemicals

Tantalum(V) tetraethoxide 2,4-pentanedionate, 99.99%, (metals basis)

CAS: 20219-33-4 | C13H27O6Ta | 460.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 039630.04
also known as 039630-04
Price (USD)
84.80
Each
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)
84.80
Each
Add to cart
Specifications
Assay Percent Notes(metals basis)
CAS20219-33-4
Chemical Name or MaterialTantalum(V) tetraethoxide 2,4-pentanedionate
Density1.5 g/mL
Flash Point>65°C (149°F)
View more
Tantalum(V) tetraethoxide 2,4-pentanedionate is used in pharmaceutical, biochemical and chemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only