Sodium hexacyanoferrate(II) decahydrate, 99%
Sodium hexacyanoferrate(II) decahydrate, 99%
Sodium hexacyanoferrate(II) decahydrate, 99%
Thermo Scientific Chemicals

Sodium hexacyanoferrate(II) decahydrate, 99%

CAS: 14434-22-1 | C6H20FeN6Na4O10 | 484.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 039660.18
also known as 039660-18
Price (USD)
66.65
Online Exclusive
74.30
Save 7.65 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
66.65
Online Exclusive
74.30
Save 7.65 (10%)
Each
Chemical Identifiers
CAS14434-22-1
Molecular FormulaC6H20FeN6Na4O10
InChI KeyJFSUDVTVQZUDOP-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[Fe++].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N
Molecular Weight (g/mol)484.06
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min
Mainly used as important raw material in pharmaceuticals, pigment, tannage, metallurgy and chemical industries. Used in producing iron blue and potassium ferricyanide, surface corrosion protecting for tannage and metal, carburization treating for carbon steel, removing iron in pharmaceutical producing. Used as oxidant, food additive, explosive and chemical reagent. Also used as additive for preventing agglomeration in melt snow in winter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only