Sulfuric acid, 50% v/v aq. soln.
Sulfuric acid, 50% v/v aq. soln.
Sulfuric acid, 50% v/v aq. soln.
Thermo Scientific Chemicals

Sulfuric acid, 50% v/v aq. soln.

CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Catalog NumberQuantity
039668.K2
also known as 039668-K2
1 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 039668.K2
also known as 039668-K2
Price (USD)
83.10
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
83.10
Each
Chemical Identifiers
CAS7664-93-9
IUPAC Namesulfuric acid
Molecular FormulaH2O4S
InChI KeyQAOWNCQODCNURD-UHFFFAOYSA-N
SMILESOS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration v/vSulfuric acid, 50% v/v aq. soln.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only