Vanadium granules, 1-3mm (0.04-0.12in), Vacuum Deposition Grade, 99.7% (metals basis)
Vanadium granules, 1-3mm (0.04-0.12in), Vacuum Deposition Grade, 99.7% (metals basis)
Vanadium granules, 1-3mm (0.04-0.12in), Vacuum Deposition Grade, 99.7% (metals basis)
Thermo Scientific Chemicals

Vanadium granules, 1-3mm (0.04-0.12in), Vacuum Deposition Grade, 99.7% (metals basis)

For Vacuum Deposition | CAS: 7440-62-2 | V | 50.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 039693.30
also known as 039693-30
Price (USD)
1,016.65
Online Exclusive
1,129.00
Save 112.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
1,016.65
Online Exclusive
1,129.00
Save 112.35 (10%)
Each
Specifications
AppearanceLight Gray granules
Assay Percent Range99.7% (Metals basis)
CAS7440-62-2
ChEBICHEBI:35170
Chemical Name or MaterialVanadium
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only