Copper gauze, 50 mesh woven from 0.23mm (0.009in) dia wire
Copper gauze, 50 mesh woven from 0.23mm (0.009in) dia wire
Copper gauze, 50 mesh woven from 0.23mm (0.009in) dia wire
Copper gauze, 50 mesh woven from 0.23mm (0.009in) dia wire
Thermo Scientific Chemicals

Copper gauze, 50 mesh woven from 0.23mm (0.009in) dia wire

CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 900 mm
500 x 900 mm
Catalog number 039713.WQ
also known as 039713-WQ
Price (USD)
224.65
Special offer
Online exclusive
Ends: 30-Jun-2026
264.00
Save 39.35 (15%)
Each
Quantity:
500 x 900 mm
Request bulk or custom format
Price (USD)
224.65
Special offer
Online exclusive
Ends: 30-Jun-2026
264.00
Save 39.35 (15%)
Each
Specifications
CAS7440-50-8
ChEBICHEBI:30052
Chemical Name or MaterialCopper gauze
EINECS Number231-159-6
IUPAC Namecopper
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only