Cobalt foil, 0.127mm (0.005in) thick, 50x50mm (2x2in), 99.9% (metals basis)
Cobalt foil, 0.127mm (0.005in) thick, 50x50mm (2x2in), 99.9% (metals basis)
Cobalt foil, 0.127mm (0.005in) thick, 50x50mm (2x2in), 99.9% (metals basis)
Thermo Scientific Chemicals

Cobalt foil, 0.127mm (0.005in) thick, 50x50mm (2x2in), 99.9% (metals basis)

CAS: 7440-48-4 | Co | 58.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 Pc.
5 Pc.
Catalog number 039715.KG
also known as 039715-KG
Price (USD)
153.65
Online Exclusive
170.00
Save 16.35 (10%)
Each
Quantity:
1 Pc.
Request bulk or custom format
Price (USD)
153.65
Online Exclusive
170.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS7440-48-4
IUPAC Namecobalt
Molecular FormulaCo
InChI KeyGUTLYIVDDKVIGB-UHFFFAOYSA-N
SMILES[Co]
View more
SpecificationsSpecification SheetSpecification Sheet
Width50mm (2in) width.
Appearance (Color)Blue silver
Assay from Supplier's CofA≥99.9% (metals basis)
FormFoil, 0.127mm (0.005in) thick.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only