Aluminum wire, 0.18mm (0.007in) dia, Puratronic™, 99.999% (metals basis)
Aluminum wire, 0.18mm (0.007in) dia, Puratronic™, 99.999% (metals basis)
Aluminum wire, 0.18mm (0.007in) dia, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Aluminum wire, 0.18mm (0.007in) dia, Puratronic™, 99.999% (metals basis)

CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 m
25 m
Catalog number 039730.G5
also known as 039730-G5
Price (USD)
83.65
Special offer
Online exclusive
Ends: 28-Dec-2026
97.90
Save 14.25
Each
Quantity:
5 m
Request bulk or custom format
Price (USD)
83.65
Special offer
Online exclusive
Ends: 28-Dec-2026
97.90
Save 14.25
Each
Specifications
Assay Percent Notes(metals basis)
CAS7429-90-5
ChEBICHEBI:33629
Chemical Name or MaterialAluminum wire
EINECS Number231-072-3
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only