Dihydrogen hexafluorotitanate, 60% w/w aq. soln.
Dihydrogen hexafluorotitanate, 60% w/w aq. soln.
Dihydrogen hexafluorotitanate, 60% w/w aq. soln.
Thermo Scientific Chemicals

Dihydrogen hexafluorotitanate, 60% w/w aq. soln.

CAS: 17439-11-1
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Catalog number 039736.A1
also known as 039736-A1
Price (USD)
156.65
Online Exclusive
174.00
Save 17.35 (10%)
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
156.65
Online Exclusive
174.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS17439-11-1
IUPAC Nametitanium(4+) dihydrogen hexafluoride
Molecular FormulaF6H2Ti
InChI KeyPFSXARRIPPWGNC-UHFFFAOYSA-J
SMILES[H+].[H+].[F-].[F-].[F-].[F-].[F-].[F-].[Ti+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration (weight/weight)60% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only